CAS RN 192863-37-9
IUPAC Name:
potassium trifluoro(thiophen-3-yl)boranuide
Synonyms:
potassium trifluoro(thiophen-3-yl)boranuide
Molecular Weight (g/mol):
190.03
Molecular Formula:
C4H3BF3KS
InChi Key:
ULOOPHJJEPUNNH-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CSC=C1
1
–
1
of
1
results
1
Promotions Available
Potassium thiophene-3-trifluoroborate, 97%, Thermo Scientific™
Potassium thiophene-3-trifluoroborate, 97%, Thermo Scientific™
CAS: 192863-37-9 Molecular Formula: C4H3BF3KS Molecular Weight (g/mol): 190.032 MDL Number: MFCD03701626 InChI Key: ULOOPHJJEPUNNH-UHFFFAOYSA-N Synonym: potassium 3-thiophenetrifluoroborate,potassium thiophene-3-trifluoroborate,potassium trifluoro thiophen-3-yl boranuide,potassium trifluoro thiophen-3-yl borate,pubchem11540,amtb173,potassiurn 3-thiophenetri tluoroborate,potassiotrifluoro 3-thienyl boron v,potassium trifluoro 3-thiophenyl boranuide,potassium trifluoro thiophen-3-yl borate 1- PubChem CID: 23677379 IUPAC Name: potassium;trifluoro(thiophen-3-yl)boranuide SMILES: [B-](C1=CSC=C1)(F)(F)F.[K+]