CAS RN 19141-40-3
IUPAC Name:
(2R)-2-phenylpropan-1-ol
Synonyms:
(2R)-2-phenylpropan-1-ol
Molecular Weight (g/mol):
136.19
Molecular Formula:
C9H12O
InChi Key:
RNDNSYIPLPAXAZ-QMMMGPOBSA-N
SMILES:
C[C@@H](CO)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(R)-(+)-2-Phenyl-1-propanol, 98+%
(R)-(+)-2-Phenyl-1-propanol, 98+%
CAS: 19141-40-3 Molecular Formula: C9H12O Molecular Weight (g/mol): 136.19 MDL Number: MFCD00145206 InChI Key: RNDNSYIPLPAXAZ-QMMMGPOBSA-N Synonym: r-2-phenyl-1-propanol,r-+-2-phenyl-1-propanol,2r-2-phenylpropan-1-ol,r-2-phenylpropanol,unii-9d7u4y35ur,2-phenyl-1-propanol, +-,2r-2-phenyl-1-propanol,+--2-phenyl-1-propanol,benzeneethanol, beta-methyl-, r,r-2-phenyl-1-propanol, +- PubChem CID: 177050 IUPAC Name: (2R)-2-phenylpropan-1-ol SMILES: C[C@@H](CO)C1=CC=CC=C1