CAS RN 190788-62-6
IUPAC Name:
8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms:
8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol):
255.12
Molecular Formula:
C15H18BNO2
InChi Key:
RNTGVJBKPQOGPS-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C2N=CC=CC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
Quinoline-8-boronic acid pinacol ester, 95%
Quinoline-8-boronic acid pinacol ester, 95%
CAS: 190788-62-6 Molecular Formula: C15H18BNO2 Molecular Weight (g/mol): 255.124 MDL Number: MFCD05663894 InChI Key: RNTGVJBKPQOGPS-UHFFFAOYSA-N Synonym: quinoline-8-boronic acid pinacol ester,8-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline,8-quinolineboronic acid pinacol ester,8-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline,amtb187,quinoline-8-boronic acid, pinacol ester PubChem CID: 17750280 IUPAC Name: 8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C=CC=N3