accessibility menu, dialog, popup

UserName

H3CNCH3OOOCH3

CAS RN 190660-97-0

IUPAC Name: methyl 4-[3-(dimethylamino)propoxy]benzoate
Synonyms: methyl 4-[3-(dimethylamino)propoxy]benzoate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: SRXQDDFKGDLAJM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OCCCN(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)