accessibility menu, dialog, popup

UserName

CH3OHO(E)

CAS RN 18999-28-5

IUPAC Name: (2E)-hept-2-enoic acid
Synonyms: (E)-2-heptenoic acid trans-acide heptene-2-oique trans-hept-2-ensaeure hept-2t-enoic acid hept-2t-ensaeure
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: YURNCBVQZBJDAJ-AATRIKPKSA-N
SMILES: CCCC\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results