CAS RN 189373-41-9
CAS RN 189373-41-9
IUPAC Name:
7-[(2-aminoethyl)amino]-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms:
7-[(2-aminoethyl)amino]-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol):
285.32
Molecular Formula:
C10H15N5O3S
InChi Key:
RDMBHRPODMXKHT-UHFFFAOYSA-N
SMILES:
CN(C)S(=O)(=O)C1=CC=C(NCCN)C2=NON=C12
1
–
1
of
1
results
1
DBD-ED [=4-(N,N-Dimethylaminosulfonyl)-7-(2-aminoethylamino)-2,1,3-benzoxadiazole], TCI America™
CAS: 189373-41-9 Molecular Formula: C10H15N5O3S Molecular Weight (g/mol): 285.322 MDL Number: MFCD01321129 InChI Key: RDMBHRPODMXKHT-UHFFFAOYSA-N Synonym: 4-(N,N-Dimethylaminosulfonyl)-7-(2-aminoethylamino)-2,1,3-benzoxadiazole PubChem CID: 4125780 IUPAC Name: 7-(2-aminoethylamino)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide SMILES: CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)NCCN