accessibility menu, dialog, popup

UserName

OO

CAS RN 1888-89-7

IUPAC Name: 2-[2-(oxiran-2-yl)ethyl]oxirane
Synonyms: 2-[2-(oxiran-2-yl)ethyl]oxirane
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: HTJFSXYVAKSPNF-UHFFFAOYNA-N
SMILES: C(CC1CO1)C1CO1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results