accessibility menu, dialog, popup

UserName

CAS RN 18877-64-0

ClH3COO(R)OHONH3CCH3CH3

CAS RN 18877-64-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results