accessibility menu, dialog, popup

UserName

CAS RN 18855-52-2

CH3OPOOH3CNHH3CCH3POOH3COH3C

CAS RN 18855-52-2

IUPAC Name: diethyl [(diethoxyphosphoryl)(dimethylazaniumyl)methyl]phosphonate
Synonyms: diethyl (diethoxyphosphoryl)(dimethylammonio)methylphosphonate
Molecular Weight (g/mol): 332.29
Molecular Formula: C11H28NO6P2
InChi Key: SQDDGOVNJMIFFT-UHFFFAOYSA-O
SMILES: CCOP(=O)(OCC)C([NH+](C)C)P(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results