accessibility menu, dialog, popup

UserName

CAS RN 18800-74-3

CH3CH3CH3OOHN(S)OONH2O

CAS RN 18800-74-3

IUPAC Name: (4S)-4-{[(tert-butoxy)carbonyl]amino}-4-carbamoylbutanoate
Synonyms: (4S)-4-[(tert-butoxycarbonyl)amino]-4-carbamoylbutanoate
Molecular Weight (g/mol): 245.26
Molecular Formula: C10H17N2O5
InChi Key: GMZBDBBBJXGPIH-LURJTMIESA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results