accessibility menu, dialog, popup

UserName

BrSO

CAS RN 18791-78-1

IUPAC Name: 4-bromothiophene-3-carbaldehyde
Synonyms: 4-bromothiophene-3-carbaldehyde
Molecular Weight (g/mol): 191.04
Molecular Formula: C5H3BrOS
InChi Key: ZDVHJQOCVBTMOF-UHFFFAOYSA-N
SMILES: BrC1=CSC=C1C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results