CAS RN 18704-67-1
IUPAC Name:
4-chloro-6-[(3Z)-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,4-dien-1-one
Synonyms:
4-chloro-6-[(3Z)-1,2-dihydropyrazol-3-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol):
194.62
Molecular Formula:
C9H7ClN2O
InChi Key:
BAWPNSUPRVQKKN-FPLPWBNLSA-N
SMILES:
ClC1=C\C(=C2\NNC=C2)C(=O)C=C1