accessibility menu, dialog, popup

UserName

FClHN

CAS RN 1868-88-8

IUPAC Name: 2-(3-chloro-4-fluorophenyl)-1H-indole
Synonyms: 2-(3-chloro-4-fluorophenyl)-1H-indole
Molecular Weight (g/mol): 245.68
Molecular Formula: C14H9ClFN
InChi Key: QNYJRNHACDLRSI-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Cl)C1=CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results