accessibility menu, dialog, popup

UserName

CAS RN 185812-86-6

IUPAC Name: bis(tri-tert-butyl-λ⁵-phosphanyl)cyclodipalladabromane
Synonyms: bis(tri-tert-butyl-λ⁵-phosphanyl)cyclodipalladabromane
Molecular Weight (g/mol): 779.31
Molecular Formula: C24H56Br2P2Pd2
InChi Key: JWWUTWDBVLEWDQ-UHFFFAOYSA-P
SMILES: CC(C)(C)P([Pd]1[Br][Pd]([Br]1)P(C(C)(C)C)(C(C)(C)C)C(C)(C)C)(C(C)(C)C)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)