accessibility menu, dialog, popup

UserName

H3CNCH3POO

CAS RN 185449-80-3

IUPAC Name: N,N-dimethyl-12,14-dioxa-13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-amine
Synonyms: N,N-dimethyl-12,14-dioxa-13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-amine
Molecular Weight (g/mol): 359.37
Molecular Formula: C22H18NO2P
InChi Key: QCHAVHXSBZARBO-UHFFFAOYSA-N
SMILES: CN(C)P1OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results