accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 18470-76-3

IUPAC Name: 3-decyloxolane-2,5-dione
Synonyms: 3-decyloxolane-2,5-dione
Molecular Weight (g/mol): 240.34
Molecular Formula: C14H24O3
InChi Key: YOWKKGPNCDIFFB-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results