accessibility menu, dialog, popup

UserName

CAS RN 18433-93-7

CH3CH3OH3CO

CAS RN 18433-93-7

IUPAC Name: 4-methyl-2-(2-methylpropyl)-1,3-dioxolane
Synonyms: 4-methyl-2-(2-methylpropyl)-1,3-dioxolane
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: PDVLTWPJDBXATJ-UHFFFAOYNA-N
SMILES: CC(C)CC1OCC(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results