accessibility menu, dialog, popup

UserName

H3CSiH3CCl

CAS RN 1833-31-4

IUPAC Name: benzyl(chloro)dimethylsilane
Synonyms: benzyl(chloro)dimethylsilane
Molecular Weight (g/mol): 184.74
Molecular Formula: C9H13ClSi
InChi Key: ABHNFDUSOVXXOA-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results