accessibility menu, dialog, popup

UserName

OHO

CAS RN 18294-87-6

IUPAC Name: 2-(cyclohex-1-en-1-yl)acetic acid
Synonyms: cyclohex-1-en-1-ylacetic acid
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: KDFBPHXESBPHTK-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CCCCC1
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results