CAS RN 18265-46-8
IUPAC Name:
disodium [(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Synonyms:
disodium D-ribofuranose 5-phosphate(2-)
Molecular Weight (g/mol):
274.07
Molecular Formula:
C5H9Na2O8P
InChi Key:
RHCIGTFSCDIJGH-CHEKPSGWSA-L
SMILES:
[Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
1
–
1
of
1
results
1
Thermo Scientific Chemicals D-Ribose-5-phosphate disodium salt hydrate, 85%
CAS: 18265-46-8 Molecular Formula: C5H9Na2O8P Molecular Weight (g/mol): 274.07 MDL Number: MFCD00135931 InChI Key: RHCIGTFSCDIJGH-CHEKPSGWSA-L Synonym: d-ribofuranose5-phosphatedisodiumsalt PubChem CID: 131875268 IUPAC Name: sodium;[(2R,3R,4R)-2,3,4-trihydroxy-5-oxopentyl] dihydrogen phosphate SMILES: [Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O