accessibility menu, dialog, popup

UserName

CAS RN 182287-51-0

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(oxan-4-yl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(oxan-4-yl)propanoate
Molecular Weight (g/mol): 272.32
Molecular Formula: C13H22NO5
InChi Key: NKYZORHKIYSSEL-SNVBAGLBSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1CCOCC1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results