CAS RN 1812-30-2
CAS RN 1812-30-2
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
CERILLIANT™ Bromazepam Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
A Certified Snap-N-Spike™ Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, and urine drug testing.