CAS RN 18039-90-2
IUPAC Name:
tris(oxolane); trichlorotitanium
Synonyms:
tris(THF); titanium(III) chloride
Molecular Weight (g/mol):
370.54
Molecular Formula:
C12H24Cl3O3Ti
InChi Key:
MIDYWIPTCYRMQF-UHFFFAOYSA-K
SMILES:
Cl[Ti](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
1
–
1
of
1
results
1
Titanium(III) chloride tetrahydrofuran complex, 97%
CAS: 18039-90-2 Molecular Formula: C12H24Cl3O3Ti Molecular Weight (g/mol): 370.56 MDL Number: MFCD01631276 InChI Key: MIDYWIPTCYRMQF-UHFFFAOYSA-K Synonym: oxolane; trichlorotitanium,titanium iii chloride tetrahydrofuran complex 1:3,trichlorotris thf titanium,titanium trichloride tri thf,trichlorotris tetrahydrofuran titanium,thf-titanium trichloride complex, 3:1,titanium trichloride tris tetrahydrofuran,trichlorotris tetrahydrofuran titanium iii,trichlorotris tetrahydrofuran tita-nium iii,titanium trichloride tetrahydrofuran complex 1:3 PubChem CID: 6097232 IUPAC Name: oxolane;trichlorotitanium SMILES: C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl