accessibility menu, dialog, popup

UserName

CAS RN 179873-36-0

IUPAC Name: methyl[(quinolin-6-yl)methyl]amine
Synonyms: methyl(quinolin-6-ylmethyl)amine
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: IIPNTNDPIZNFRU-UHFFFAOYSA-N
SMILES: CNCC1=CC=C2N=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results