accessibility menu, dialog, popup

UserName

CAS RN 17966-84-6

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHLu

CAS RN 17966-84-6

IUPAC Name: lutetium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: lutetium(3+) triacetylacetonate
Molecular Weight (g/mol): 472.29
Molecular Formula: C15H21LuO6
InChi Key: HXFHOORFYPCGNF-UHFFFAOYSA-N
SMILES: [Lu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results