accessibility menu, dialog, popup

UserName

H3COPOPOOCH3OCH3OCH3

CAS RN 17919-34-5

IUPAC Name: diethyl ({4'-[(diethoxyphosphoryl)methyl]-[1,1'-biphenyl]-4-yl}methyl)phosphonate
Synonyms: diethyl {4'-[(diethoxyphosphoryl)methyl]-[1,1'-biphenyl]-4-yl}methylphosphonate
Molecular Weight (g/mol): 454.44
Molecular Formula: C22H32O6P2
InChi Key: DRCOGQJUVZRNSQ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(C=C1)C1=CC=C(CP(=O)(OCC)OCC)C=C1)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results