accessibility menu, dialog, popup

UserName

CH3SiH3CCH3S

CAS RN 17873-08-4

IUPAC Name: trimethyl[(phenylsulfanyl)methyl]silane
Synonyms: trimethyl[(phenylsulfanyl)methyl]silane
Molecular Weight (g/mol): 196.38
Molecular Formula: C10H16SSi
InChi Key: UOQDIMYVQSHALM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CSC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results