accessibility menu, dialog, popup

UserName

ClHN

CAS RN 17784-47-3

IUPAC Name: acridin-10-ium chloride
Synonyms: acridin-10-ium chloride
Molecular Weight (g/mol): 215.68
Molecular Formula: C13H10ClN
InChi Key: XUESTGHCVFYOLL-UHFFFAOYSA-N
SMILES: [Cl-].C1=CC2=CC3=CC=CC=C3[NH+]=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results