accessibility menu, dialog, popup

UserName

CAS RN 177735-09-0

IUPAC Name: (3-methylthiophen-2-yl)boronic acid
Synonyms: 3-methylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 141.98
Molecular Formula: C5H7BO2S
InChi Key: MOOOGTOEKZWEJA-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results