CAS RN 1772-43-6
CAS RN 1772-43-6
IUPAC Name:
2,4,4-trimethyl-4,5-dihydro-1,3-oxazole
Synonyms:
2,4,4-trimethyl-5H-1,3-oxazole
Molecular Weight (g/mol):
113.16
Molecular Formula:
C6H11NO
InChi Key:
HZRZMHNRCSIQFT-UHFFFAOYSA-N
SMILES:
CC1=NC(C)(C)CO1
1
–
1
of
1
results
1
2,4,4-Trimethyl-2-oxazoline 98.0+%, TCI America™
CAS: 1772-43-6 Molecular Formula: C6H11NO Molecular Weight (g/mol): 113.16 MDL Number: MFCD00005301 InChI Key: HZRZMHNRCSIQFT-UHFFFAOYSA-N Synonym: 2,4,4-trimethyl-2-oxazoline,2,4,4-trimethyl-4,5-dihydrooxazole,4,5-dihydro-2,4,4-trimethyloxazole,oxazole, 4,5-dihydro-2,4,4-trimethyl,2,4,4-trimethyloxazoline,oxazole,4,5-dihydro-2,4,4-trimethyl,2,4,4-trimethyl-1,3-oxazoline,pubchem24351,acmc-1bsc7,2,4,4-trimethyl-5h-oxazole PubChem CID: 74496 IUPAC Name: 2,4,4-trimethyl-5H-1,3-oxazole SMILES: CC1=NC(CO1)(C)C