accessibility menu, dialog, popup

UserName

CAS RN 1771-18-2

IUPAC Name: 2-methoxy-10H-phenothiazine
Synonyms: 2-methoxy-10H-phenothiazine
Molecular Weight (g/mol): 229.3
Molecular Formula: C13H11NOS
InChi Key: DLYKFPHPBCTAKD-UHFFFAOYSA-N
SMILES: COC1=CC=C2SC3=CC=CC=C3NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results