accessibility menu, dialog, popup

UserName

NH2NN

CAS RN 17635-45-9

IUPAC Name: 4-(1H-pyrazol-1-yl)aniline
Synonyms: 4-(pyrazol-1-yl)aniline
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: CSFIQHZIFKIQNO-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)