accessibility menu, dialog, popup

UserName

CAS RN 17572-97-3

KKKHOONNOOOOOO

CAS RN 17572-97-3

IUPAC Name: tripotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate
Synonyms: tripotassium edta
Molecular Weight (g/mol): 406.52
Molecular Formula: C10H13K3N2O8
InChi Key: FYZXEMANQYHCFX-UHFFFAOYSA-K
SMILES: [K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results