accessibility menu, dialog, popup

UserName

CAS RN 175277-98-2

IUPAC Name: 2,4-dichloro-6-methylbenzonitrile
Synonyms: 2,4-dichloro-6-methylbenzonitrile
Molecular Weight (g/mol): 186.04
Molecular Formula: C8H5Cl2N
InChi Key: NFPYAMXNKNDVDS-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(Cl)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results