accessibility menu, dialog, popup

UserName

CH3NNNH2

CAS RN 175202-37-6

IUPAC Name: 3-(1-methyl-1H-pyrazol-3-yl)aniline
Synonyms: 3-(1-methylpyrazol-3-yl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: ZRXOZSROZJGRRH-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(N)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)