accessibility menu, dialog, popup

UserName

CAS RN 175201-51-1

IUPAC Name: (7R)-4,7-dimethyl-4H,7H-pyrazolo[1,5-a]pyrimidine-3-carboxylate
Synonyms: (7R)-4,7-dimethyl-7H-pyrazolo[1,5-a]pyrimidine-3-carboxylate
Molecular Weight (g/mol): 192.2
Molecular Formula: C9H10N3O2
InChi Key: NDSVSMFAGULPFP-ZCFIWIBFSA-M
SMILES: C[C@@H]1C=CN(C)C2=C(C=NN12)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results