accessibility menu, dialog, popup

UserName

CH3OOSOOH3C

CAS RN 175172-61-9

IUPAC Name: 2-methoxyethyl methanesulfonate
Synonyms: 2-methoxyethyl methanesulfonate
Molecular Weight (g/mol): 154.18
Molecular Formula: C4H10O4S
InChi Key: BCKAHDGFNHDQST-UHFFFAOYSA-N
SMILES: COCCOS(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results