accessibility menu, dialog, popup

UserName

H3COBrCH3H3CH3CCH3

CAS RN 175136-57-9

IUPAC Name: 2-bromo-1-(3-ethyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(3-ethyl-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 337.3
Molecular Formula: C18H25BrO
InChi Key: BBHJYKJAWKZSHS-UHFFFAOYSA-N
SMILES: CCC1=CC2=C(C=C1C(=O)CBr)C(C)(C)CCC2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)