CAS RN 174125-30-5
IUPAC Name:
(5-chloropent-1-yn-1-yl)triethylsilane
Synonyms:
(5-chloropent-1-yn-1-yl)triethylsilane
Molecular Weight (g/mol):
216.82
Molecular Formula:
C11H21ClSi
InChi Key:
YOFYXFCOGXKCGG-UHFFFAOYSA-N
SMILES:
CC[Si](CC)(CC)C#CCCCCl
1
–
1
of
1
results
1
Promotions Available
1-Chloro-5-triethylsilyl-4-pentyne, 97%
1-Chloro-5-triethylsilyl-4-pentyne, 97%
CAS: 174125-30-5 Molecular Formula: C11H21ClSi Molecular Weight (g/mol): 216.82 MDL Number: MFCD00671352 InChI Key: YOFYXFCOGXKCGG-UHFFFAOYSA-N Synonym: 1-chloro-5-triethylsilyl-4-pentyne,5-chloropent-1-ynyl triethyl silane,5-chloropent-1-ynyl triethylsilane,1-triethylsilyl-5-chloro-1-pentyne,5-chloranylpent-1-ynyl triethyl silane,5-chloropent-1-yn-1-yl triethylsilane,5-chloropent-1-yn-1-yl triethyl silane,silane, 5-chloro-1-pentyn-1-yl triethyl PubChem CID: 2757930 IUPAC Name: 5-chloropent-1-ynyl(triethyl)silane SMILES: CC[Si](CC)(CC)C#CCCCCl