accessibility menu, dialog, popup

UserName

CAS RN 17289-26-8

IUPAC Name: 1-methyl-1H-imidazole-4-carbaldehyde
Synonyms: 1-methylimidazole-4-carbaldehyde
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: CQZXDIHVSPZIGF-UHFFFAOYSA-N
SMILES: CN1C=NC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results