CAS RN 17289-20-2
IUPAC Name:
methyl 1-methyl-1H-imidazole-5-carboxylate
Synonyms:
methyl 3-methylimidazole-4-carboxylate
Molecular Weight (g/mol):
140.14
Molecular Formula:
C6H8N2O2
InChi Key:
AKDPLDCXQNEMCL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CN=CN1C
1
–
1
of
1
results
1
Methyl 1-methylimidazole-5-carboxylate, 98%, Thermo Scientific™
CAS: 17289-20-2 Molecular Formula: C6H8N2O2 Molecular Weight (g/mol): 140.14 MDL Number: MFCD01567300 InChI Key: AKDPLDCXQNEMCL-UHFFFAOYSA-N Synonym: methyl 1-methyl-1h-imidazole-5-carboxylate,methyl 1-methylimidazole-5-carboxylate,methyl1-methyl-1h-imidazole-5-carboxylate,3-methyl-3h-imidazole-4-carboxylic acid methyl ester,1h-imidazole-5-carboxylic acid, 1-methyl-, methyl ester,1-methyl-1h-imidazole-5-carboxylic acid methyl ester,pubchem9004,acmc-209e5h,1-methylimidazole-5-carboxylic acid methyl ester,1h-imidazole-5-carboxylicacid, 1-methyl-, methyl ester PubChem CID: 2736896 SMILES: COC(=O)C1=CN=CN1C