accessibility menu, dialog, popup

UserName

CAS RN 172514-63-5

IUPAC Name: 2-bromo-5-octylthiophene
Synonyms: 2-bromo-5-octylthiophene
Molecular Weight (g/mol): 275.25
Molecular Formula: C12H19BrS
InChi Key: USLMRHXOAXBGCS-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(Br)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results