accessibility menu, dialog, popup

UserName

CAS RN 17193-40-7

IUPAC Name: methyl 2-amino-3-(4-nitrophenyl)propanoate hydrochloride
Synonyms: methyl 4-nitrophenylalaninate hydrochloride
Molecular Weight (g/mol): 260.67
Molecular Formula: C10H13ClN2O4
InChi Key: BTHMRXRBXYHLRA-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results