CAS RN 17129-06-5
IUPAC Name:
4-ethoxy-1,1,1-trifluorobut-3-en-2-one
Synonyms:
4-ethoxy-1,1,1-trifluorobut-3-en-2-one
Molecular Weight (g/mol):
168.12
Molecular Formula:
C6H7F3O2
InChi Key:
YKYIFUROKBDHCY-UHFFFAOYSA-N
SMILES:
CCOC=CC(=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 97%
4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 97%
CAS: 17129-06-5 Molecular Formula: C6H7F3O2 Molecular Weight (g/mol): 168.12 MDL Number: MFCD00192131 InChI Key: YKYIFUROKBDHCY-UHFFFAOYSA-N Synonym: 4-ethoxy-1,1,1-trifluoro-3-buten-2-one,4-ethoxy-1,1,1-trifluorobut-3-en-2-one,e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one,3e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one,etfbo,2-ethoxyvinyl trifluoromethyl ketone,e-4-ethoxy-1,1,1-trifluoro-3-buten-2-one,3-buten-2-one, 4-ethoxy-1,1,1-trifluoro,4-ethoxy,4-ethoxy-1,1,1-trifluoro-but-3-en-2-one PubChem CID: 5709222 IUPAC Name: (E)-4-ethoxy-1,1,1-trifluorobut-3-en-2-one SMILES: CCOC=CC(=O)C(F)(F)F