accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOOH3CNH2HCl

CAS RN 171110-72-8

IUPAC Name: 1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Synonyms: 1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Molecular Weight (g/mol): 280.75
Molecular Formula: C11H21ClN2O4
InChi Key: KFYCQKLSGMAVQH-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C1CC(N)CN1C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)