accessibility menu, dialog, popup

UserName

CAS RN 16986-24-6

A(a)

CAS RN 16986-24-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 144.25
Molecular Formula: C2H12B10
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results