accessibility menu, dialog, popup

UserName

CH3SiH3CCH3FFSiCH3H3CCH3FF

CAS RN 16956-91-5

IUPAC Name: trimethyl[2,3,5,6-tetrafluoro-4-(trimethylsilyl)phenyl]silane
Synonyms: trimethyl[2,3,5,6-tetrafluoro-4-(trimethylsilyl)phenyl]silane
Molecular Weight (g/mol): 294.44
Molecular Formula: C12H18F4Si2
InChi Key: RIGHSQASRZUCPK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=C(F)C(F)=C(C(F)=C1F)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results