accessibility menu, dialog, popup

UserName

CAS RN 1694-09-3

NaH3CNSOOOCNCH3H3CNCH3SOOO

CAS RN 1694-09-3

IUPAC Name: sodium [4-(dimethylamino)phenyl]bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Synonyms: sodium [4-(dimethylamino)phenyl]bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Molecular Weight (g/mol): 733.87
Molecular Formula: C39H40N3NaO6S2
InChi Key: AXMCIYLNKNGNOT-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results