accessibility menu, dialog, popup

UserName

CH3NCH3CH3CH3I

CAS RN 16829-91-7

IUPAC Name: tetraoctylazanium iodide
Synonyms: tetraoctylazanium iodide
Molecular Weight (g/mol): 593.81
Molecular Formula: C32H68IN
InChi Key: KGPZZJZTFHCXNK-UHFFFAOYSA-M
SMILES: [I-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results