accessibility menu, dialog, popup

UserName

CH3NCH3ONNNCH3CH3

CAS RN 1679-98-7

IUPAC Name: 4-{5-[4-(diethylamino)phenyl]-1,3,4-oxadiazol-2-yl}-N,N-diethylaniline
Synonyms: 4-{5-[4-(diethylamino)phenyl]-1,3,4-oxadiazol-2-yl}-N,N-diethylaniline
Molecular Weight (g/mol): 364.49
Molecular Formula: C22H28N4O
InChi Key: UZGVMZRBRRYLIP-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C1=NN=C(O1)C1=CC=C(C=C1)N(CC)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results